Whole Allergen
t23 Italian/Mediterranean/Funeral cypress Pollen
t23 Italian/Mediterranean/Funeral cypress Pollen Scientific Information
Type:
Whole Allergen
Display Name:
Italian/Mediterranean/Funeral cypress Pollen
Allergen code:
t23
Family:
Cupressaceae
Species:
Cupressus sempervirens
Route of Exposure:
Inhalation
Source Material:
Pollen
Latin Name:
Cupressus sempervirens
Other Names:
Common cypress, funeral cypress, Italian cypress, evergreen cypress, graveyard cypress, Tuscan cypress
WHO/ICD-11 code:
XM4WW3
(ICD-11 is currently under implementation by WHO and the ICD-11 codes displayed in the encyclopedia may not yet be available in all countries)
Summary
Italian cypress (Cupressus sempervirens), also named as Mediterranean or funeral cypress is a traditional, ornamental tree often used in cemeteries. This evergreen, wind-pollinated, monecious tree can grow up to a height of 35-40 m and is capable of producing pollens in huge quantities. This tree is drought and heat tolerant and is commonly seen in areas, such as farmlands and forests. The tree is commonly found in the US, Europe, Japan, Australia, North-East African countries and Middle-Eastern countries. The prevalence rate of cypress pollen allergy has been reported between 0.6% and 32%, according to the studies conducted in various geographical region. Exposure to Italian cypress tree pollen can induce symptoms, such as allergic rhinitis, rhino-conjunctivitis, asthma, and contact dermatitis in rare cases. Cup s 1 and Cup s 3 have been identified as the major allergens in the Italian cypress tree pollen. Studies have demonstrated high cross-reactivity between pollens from Italian cypress and other related species, such as Arizona cypress, mountain juniper, Japanese cypress, and Japanese cedar. This species pollen has also exhibited cross-reactivity with scrub cypress pine, African fern pine and peach. 
Allergen
Nature
Italian cypress (Cupressus sempervirens), also named as Mediterranean or funeral cypress (1) is a traditional ornamental tree, widely used in cemeteries, due to its conveniently contained roots  (2, 3). This evergreen, wind-pollinated, monecious tree can attain a height ranging between 35 to 40 m, with the diameter of trunk being 1 m in thickness. The leaves are dark green and scaly textured, whereas the bark is greyish brown in colour with thin rib-like structure. The flowers of these trees are unisexual, with male flowers being yellow coloured, cylindrical-shaped, and 3-5 mm in length, whereas the female flowers are brownish green, and spherical in shape. The female flowers after ripening (~1 year), convert into brownish grey, oval-shaped woody cones that are 2-4 cm in length. The pollination period generally occurs between mid-winter and early spring (2). However, it may extend until late April or even May in Mediterranean areas. These trees generally produce large quantities of pollen grains, with a pollen output of about 276,000 million per tree (3). Branches and cones from young Italian cypress tree are used for medicinal and pharmacological purposes, such as antidiabetic, antiseptic, anticancer and other treatments (4).   
Habitat
Italian cypress grow at varied altitudes and on different areas, such as farmland and forests   (3). The natural environment required for this tree to grow well is either in the Mediterranean climate, i.e. hot, dry summers and wet winters or semi-arid, steppe climates in the eastern range (2). Regions where winters have warm climate and comparatively higher rainfall, increased pollen release from the cypress trees are seen, leading to higher allergy cases (5). The tree is tolerant to heat and drought, and requires light and an annual rainfall of ~200 mm for growth. A rich, moist soil with good aeration, and having a neutral pH is most favourable for the growth of this tree (2). 
Taxonomy
Cupressaceae is a conifer family, that is categorized into seven different subfamilies, 30 genera and close to 160 species. Italian cypress is a tree that belongs to the family of Cupressaceae and genus Cupressus. Interestingly, this genus Cupressus, along with the four other genera, i.e. Chamaecyparis, Hesperocyparis, Juniperus and Cryptomeria are considered to be the most essential taxa for allergic diseases (3). 
Taxonomic tree of  Italian Cypress (6, 7)
Domain Eukaryota
Kingdom Plantae
Phylum Spermatophyta
Subphylum Gymnospermae
Class Pinopsida
Order Pinales
Family Cupressaceae
Genus Cupressus
Species Cupressus sempervirens
Tissue
All tree pollens belonging to Cupressaceae family (including Italian cypress) possess spheroidal-shaped pollen grains, with the size of each grain ranging from 25 to 40 µm (3). This was evidenced in a study that determined the pollen structure of Italian cypress (Cupressus sempervirens), where the pollen grain was found to be spheroidal and polar in shape and the diameter of pollen ranged between 25.18 µm to 32.5 µm. However, since the pollen grains lacked apertures, they displayed fake pores of size 10 µm (minor axis) x 15 µm (major axis) on the surface (8).
Cup s 1 and Cup s 3 are identified as the two most potential allergens from the pollens of Italian cypress (1, 9) .
Epidemiology
Worldwide distribution
Pollens from Italian cypress trees are considered as a primary contributor to seasonal allergic rhinitis (AR) in the Mediterranean areas (10). It has been reported that almost half of the total pollination level is accounted by this pollinating species in this region. Cypress pollinosis is represented by asthma, conjunctivitis, and AR (3).
Wide variation in the prevalence rate of allergy to cypress pollen has been reported in the studies mentioned below, that ranged from 0.6% to 32%. The prevalence rate in these studies was identified, either by conducting questionnaire surveys or through SPT results.
In a cross-sectional study conducted in France, the prevalence of cypress pollen allergy was reported between 0.6%-2.4%, among patients with AR (hay fever) (11).
Another study conducted in Greece reported 12.7% (166 out of 1311) of patients with respiratory allergy, being sensitized to cypress pollen (12).
Furthermore, in a study conducted in Israel, a relatively higher prevalence (24%- 32%) of sensitization to cypress pollen was reported in patients suffering from AR (13).
A retrospective study was conducted in Italy among 1397 patients who complained of having conjunctival or respiratory disorders between the year 1995 and 1998. The results showed an annual increase in the rate of sensitization to Italian cypress pollen, that ranged from 7.2% to 22% between these years (14).
In a study conducted among school-going children (6 to 17 years) in Greece, the prevalence rate of sensitization to various pollens was determined. Among 675 children who participated in the study, 231 (34.2%) were diagnosed as having AR. Of 231 patients, 4.8% (11 out of 231) of patients reported sensitization to cypress pollen. Interestingly, children aged 6 to 11 years showed higher sensitization to these pollens in comparison to children aged 12 to 17 years (6-11 years: 63.6% vs 12-17 years: 36.4%). Furthermore, a significant co-sensitization was seen between pine and cypress allergens (p=0.006) (15). 
Environmental Characteristics
Worldwide distribution
Italian cypress tree is considered by various sources as a native species of Turkey, the Aegean Islands, North-East African countries, i.e., Libya and Tunisia, Cyprus, and some Middle Eastern countries, namely Syria, Jordan, Lebanon, and Iran. Natural stands are found in the south-west part of Mediterranean region (2). Furthermore, cypress pollinosis is observed worldwide in several countries, such as the US, Europe, South Africa, Iran, Australia, Japan and the Mediterranean region (3).
Route of Exposure
Main
One of the significant routes of exposure to Italian cypress pollen is through airway inhalation. Pollens from Italian cypress trees are wind-pollinated and can spread across large distances. The pollen grains are considered as the carriers of allergens, since they contain small particles, known as orbicules on their surface, that are responsible for containing these allergens (3).
Secondary
Direct skin contact is another route of exposure. This was evidenced in a case study, where a 56-year-old woman, after coming in contact with the resin of Italian cypress leaves, developed contact dermatitis (7). 
Clinical Relevance
Cypress pollen hypersensitivity is a significant contributor to pollinosis, particularly in countries of Mediterranean region (16). Cypress pollinosis is responsible for inducing allergic symptoms, such as rhino-conjunctivitis, AR, and asthma in sensitive individuals (17). Genetic susceptibility, elevated pollen exposure and past experience of cypress allergy are considerable risk factors for determining allergic and atopic patients. Interestingly, AR is the most frequently observed symptom, whereas rhino-conjunctivitis is the most debilitating symptom of cypress pollen allergy (3). 
Allergic rhinitis (AR) and allergic rhino-conjunctivitis (ARC)
A steady rise in the prevalence of allergic conditions, such as AR, rhino-conjunctivitis and asthma has been observed in elderly patients sensitized to allergens. In a case-control study conducted in Italy, elderly patients (>65 years) with AR and/or rhino-conjunctivitis (cases) were compared to young adults (15 – 40 years) who acted as control. According to the results, among all the allergens extracts analysed by SPT, cypress pollen was the most common sensitizing allergen in the elderly (17).
In a study performed in Turkey on 455 patients diagnosed with AR and/or asthma, 81.3% (370 out of 455) of patients had AR, and 18.7% (85/455) of patients had asthma as well as AR. Out of these patients, 14.3% (65 out of 455) were found sensitized to cypress pollen  (18).
In a study conducted on 47 cypress pollen-allergic patients in France, an association between cypress pollen concentration levels, rhino-conjunctivitis symptoms (itching, redness, teary eyes, sneezing, runny/stuffed nose, wheezing, cough or asthma) and quality of life (QoL) was reported. According to the study, a logarithmic curve was reported between cypress pollen count and rhino-conjunctivitis symptoms. This evidence was supported by the results that revealed a remarkable elevation in rhino-conjunctivitis symptoms, when the pollen count increased from 0 to 500 grains/m3. However a gradual decrease in these symptoms were noted, in spite of an exposure to increased number of pollen grains (>500 grains/m3), displaying a plateau effect at increased level of pollen exposure. Furthermore, a linear association was observed between cypress pollen count and QoL score, which indicated that, with every increase in 500 grains of cypress pollen, an increase in QoL score was reported. The study, however could not identify the threshold level for the occurrence of allergic symptoms or QoL in these allergic individuals, indicating larger studies to be conducted for determining this potential threshold (19). 
A retrospective study was conducted over a period of 4 years in Italy on 1397 individuals complaining of respiratory tract or conjunctival disease. According to the results, the prevalence of positive SPT in patients allergic to Italian cypress pollen was found to be 17.4% (243 out of 1397). Among these individuals, 64.6% (157 out of 243) had rhino-conjunctivitis, 27.2% (66 out of 243) had asthma and 8.2% (20 out of 243) had both asthma as well as rhino-conjunctivitis (14).
Contact dermatitis
A case of a 56-year-old woman in France who suffered lesions on her body, after coming in contact with the resin of Italian cypress leaves. The patient had a past history of developing a skin response towards adhesive bandages. The patch test performed showed positive association between the leaves, cone, resin of the tree species and colophonium derivates used in adhesive bandages samples. This concluded that contact dermatitis developed in the patient could be a result of cross-reactivity between colophonium and Italian cypress allergen (7). 
Prevention and Therapy
Allergen immunotherapy
A double-blind, placebo-controlled study was conducted among 40 patients, where 20 patients with rhino-conjunctivitis and cypress pollen allergy were administered with either sublingual-swallow immunotherapy (SLIT; n=10) or subcutaneous immunotherapy (SCIT; n=10), using Juniperus ashei extract and 20 patients were administered placebo. According to the results, treatment with SLIT and SCIT showed good improvement in the clinical symptoms as well as decrease in the inflammation parameters (serum levels of eosinophilic cationic protein and nasal levels) in patients with rhino-conjunctivitis due to cypress allergy (20). 
Prevention strategies
Avoidance
Italian cypress pollen allergy can be prevented or reduced by using a few precautionary measures, such as washing hair before sleeping, using protective sunglasses while stepping out of the house, restricting oneself from doing exercise outside the home, keeping doors and windows closed, and avoiding drying of linen clothes outside (3).
Furthermore, to minimize the amount of pollen into respiratory system, it is suggested to either stay indoors or wear a mask, while moving outdoors. If the patient’s health condition deteriorates, it is further advised to move to an area, where there is less pollination (21).
Molecular Aspects
Allergenic molecules
The World Health Organization (WHO) and International Union of Immunological Societies’ (IUIS) Allergen Nomenclature Subcommittee have registered four allergenic components, i.e., Cup s 1, Cup s 2, Cup s 3 and Cup s 7 for Italian cypress pollen (9).
Allergen Biochemical Name Molecular Weight (kDa) Allergenicity
Cup s 1 Pectate lyase 43
  • Major allergen
  • A study conducted on 187 patients (2-14 years) with allergic rhino-conjunctivitis and/or asthma, 20.9% (39 out of 187) of patients showed specific-IgE (sIgE) reactivity towards Cup s 1 (22).
  • Another study showed that, out of 30 cypress-pollen sensitized patients, 22 (73.3%) showed positive IgE-response towards Cup s 1 (23).
  • High sequence similarity was noted between Cup s 1 and other cypress pollen allergens (Cry j 1, Cha o 1, Jun o 1, Jun v 1, Jun a 1, Cup a 1) (1).
Cup s 2 Polygalacturonase 43
  • In a study conducted among 30 cypress-pollen allergic patients, 21 (70%) were found sensitized towards Cup s 2 (23).
Cup s 3 Thaumatin-like protein 34
  • Major allergen
  • In a study conducted on sera of 23 Italian cypress–allergic patients, IgE reactivity towards Cup s 3 was observed in 69.6% (16 of 23) of the patients (10).
  • ~95% sequence similarity was observed between Cup s 3 and Jun a 3 (24)
Cup s 7 Gibberellin-regulated protein (GRP) 9.5
  • Five individuals sensitized to Pru p 7, Cry j 1 and Cup a 1, showed IgE-reactivity towards Cup s 7 allergen (25). 
Biomarkers of severity
Cup s 1 is considered as a major allergen of Italian cypress pollen (C. sempervirens) with a study displaying specific immunoglobulin (Ig)E reactivity of 20.9% towards Cup s 1 (22). Where another study involving 30 cypress-pollen sensitised patients, 73.3% (n=22) showed positive IgE-response towards Cup s 1 allergen (23).
Cup s 3, another major allergen has been identified in Italian cypress pollen. This allergen was recognized on the basis of cross-reactivity and sequence homology with other cypress pollen allergens, despite exhibiting low protein expressions. Variations with respect to content of Cup s 3 allergen in pollens from various regions should be taken into consideration, while preparing extracts for diagnostic purposes (24).
Of interest, Cup a 1 from Cupressus Arizonica pollen has been identified as a genuine marker allergen for sensitization to cypress pollen (25). 
Cross-reactivity
Italian cypress pollen has shown to cross-react and share high sequence homology with closely-related species, such as Arizona cypress, mountain juniper and Japanese cypress, belonging to the same family Cupressaceae and Japanese cedar which belongs to the same class Pinopsida as Italian Cypress (26).
High degree of sequence similarity i.e. 95.1% has been reported between Cup s 1 (Cupressus sempervirens) and Cup a 1 (Cupressus Arizonica) (27). Furthermore, Cup a 1 has shown to exhibit high sequence homology of 91% with Jun a 1 (Juniperus ashei), 75% with Cry j 1 (Cryptomeria japonica) and 81% with Cha o 1 (Chamaecyparis obtuse) (26, 28). These major allergens from the same family and class, are highly cross-reactive, as they share between 70% to 95% of sequence similarity (26).
Another major allergen from C. sempervirens species, Cup s 3 has revealed to have 95% sequence homology and strong cross-reactivity with Jun a 3 (J. ashei) (24).
Limited cross-reactivity was displayed between Italian cypress pollen, scrub cypress pine (Callitris verrucosa) and African fern pine (Podocarpus gracilior) (29).
Olive pollen has shown to cross-react with cypress pollen, pine pollen, mugwort, and birch in a study, where IgE binding to these pollen extracts was completely inhibited by extract of olive pollen. This showed that proteins of olive pollens share allergenic epitopes that are in common with the proteins of the other pollen extracts, and hence the cross-reactivity (30).
Cross-reactivity through IgE binding has been demonstrated among Italian cypress, white cypress pine, rye grass, ragweed, olive, birch, couch grass, plantain, lamb’s quarters, cocksfoot and wall pellitory (31).
Cross-reactivity has also been reported between peach and cypress pollen. This was evidenced in a case series of seven cypress pollen-allergic patients, who immediately after consuming peach developed OAS, urticaria or Quincke edema (32).
Compiled By
Author: Turacoz Healthcare Solutions
Reviewer: Dr. Fabio Iachetti
Last reviewed: January 2021
References
  1. Shahali Y, Sutra J-P, Peltre G, Charpin D, Sénéchal H, Poncet P. IgE Reactivity to Common Cypress (C. Sempervirens) Pollen Extracts: Evidence for Novel Allergens. The World Allergy Organization journal. 2010;3:229-34.
  2. Caudullo G, De Rigo D. Cupressus sempervirens in Europe: distribution, habitat, usage and threats. European Atlas of Forest Tree Species Publications Office of the European Union, Luxembourg pp e015be7. 2016.
  3. Charpin D, Pichot C, Belmonte J, Sutra J-P, Zidkova J, Chanez P, et al. Cypress Pollinosis: from Tree to Clinic. Clinical Reviews in Allergy & Immunology. 2017;56.
  4. Hussain MK, Saquib M, Ahamad T, Khatoon S, Khan MF. Mediterranean Cypress “Cupressus sempervirens”: A Review on Phytochemical and Pharmacological Properties. Current Traditional Medicine. 2019;5(4):278-97.
  5. Monroy-Colín A, Manzano JMM, Silva-Palacios I, Tormo-Molina R, Pecero-Casimiro R, Gonzalo-Garijo Á, et al. Phenology of Cupressaceae urban infrastructure related to its pollen content and meteorological variables. Aerobiologia. 2020:1-21.
  6. CABI. Invasive Species Compendium. 2019.
  7. Samaran Q, Clark E, Dereure O, Raison‐Peyron N. Allergic contact dermatitis to Cupressus sempervirens resin and cross‐reaction with colophonium. Contact Dermatitis. 2020.
  8. Şahin AA, Aslım B, Tan S, Alan Ş, Pınar NM. Differences in structure, allergenic protein content and pectate lyase enzyme activity of some Cupressaceae pollen. Turkish Journal of Biochemistry. 2018;43(4):435-46.
  9. WHO/IUIS. Cupressus sempervirens (Common cypress) 2019 [27.12.2020]. Available from: http://www.allergen.org/search.php?allergensource=Cupressus+sempervirens.
  10. Togawa A, Panzani RC, Garza MA, Kishikawa R, Goldblum RM, Midoro-Horiuti T. Identification of italian cypress (Cupressus sempervirens) pollen allergen Cup s 3 using homology and cross-reactivity. Ann Allergy Asthma Immunol. 2006;97(3):336-42.
  11. Charpin D, Hughes B, Mallea M, Sutra JP, Balansard G, Vervloet D. Seasonal allergic symptoms and their relation to pollen exposure in south‐east France. Clinical & Experimental Allergy. 1993;23(5):435-9.
  12. Gioulekas D, Papakosta D, Damialis A, Spieksma F, Giouleka P, Patakas D. Allergenic pollen records (15 years) and sensitization in patients with respiratory allergy in Thessaloniki, Greece. Allergy. 2004;59(2):174-84.
  13. Geller-Bernstein C, Waisel Y, Lahoz C. Environment and sensitization to cypress in Israel. Allerg Immunol (Paris). 2000;32(3):92-3.
  14. Papa G, Romano A, Quaratino D, Di Fonso M, Viola M, Cristina Artesani M, et al. Prevalence of sensitization to Cupressus sempervirens: a 4-year retrospective study. Science of The Total Environment. 2001;270(1):83-7.
  15. Katotomichelakis M, Danielides G, Iliou T, Anastassopoulos G, Nikolaidis C, Kirodymos E, et al. Allergic sensitization prevalence in a children and adolescent population of northeastern Greece region. International journal of pediatric otorhinolaryngology. 2016;89:33-7.
  16. Di Rienzo V, Pucci S, D'Alò S, Di Cara G, Incorvaia C, Frati F, et al. Effects of high-dose sublingual immunotherapy on quality of life in patients with cypress-induced rhinitis: a placebo-controlled study. Clinical & Experimental Allergy Reviews. 2006;6(3):67-70.
  17. Ventura MT, Gelardi M, D'Amato A, Buquicchio R, Tummolo R, Misciagna G, et al. Clinical and cytologic characteristics of allergic rhinitis in elderly patients. Ann Allergy Asthma Immunol. 2012;108(3):141-4.
  18. Sin AZ, Ersoy R, Gulbahar O, Ardeniz O, Gokmen NM, Kokuludag A. Prevalence of cypress pollen sensitization and its clinical importance in Izmir, Turkey, with cypress allergy assessed by nasal provocation. J Investig Allergol Clin Immunol. 2008;18(1):46-51.
  19. Rakotozandry T, Cassagne E, Martin S, Alauzet P, Navarro I, Delcroux C, et al. Exposure to Cypress Pollens and Subsequent Symptoms: A Panel Study. International archives of allergy and immunology. 2019;180(2):135-41.
  20. Ventura MT, Carretta A, Tummolo RA, Buquicchio R, Arsieni A, Murgia N. Clinical data and inflammation parameters in patients with cypress allergy treated with sublingual swallow therapy and subcutaneous immunotherapy. Int J Immunopathol Pharmacol. 2009;22(2):403-13.
  21. Xie Z-J, Guan K, Yin J. Advances in the clinical and mechanism research of pollen induced seasonal allergic Asthma. American journal of clinical and experimental immunology. 2019;8(1):1.
  22. Feliu A, Gonzalez-de-Olano D, Gonzalez E, Rodriguez B, Ruiz-Hornillos J, Jimeno L, et al. A Multicenter Study of Sensitization Profiles in an Allergic Pediatric Population in an Area With High Allergen Exposure. J Investig Allergol Clin Immunol. 2013;23(5):337-44.
  23. Shahali Y, Sutra JP, Haddad I, Vinh J, Guilloux L, Peltre G, et al. Proteomics of cypress pollen allergens using double and triple one‐dimensional electrophoresis. Electrophoresis. 2012;33(3):462-9.
  24. Togawa A, Panzani R, Garza M, Goldblum R, Midoro-Horiuti T. Isolation and characterization of the major allergen from Italian cypress (Cupressus sempervirens) pollen. Journal of Allergy and Clinical Immunology. 2004;113(2):S300.
  25. Tuppo L, Alessandri C, Giangrieco I, Ciancamerla M, Rafaiani C, Tamburrini M, et al. Isolation of cypress gibberellin-regulated protein: Analysis of its structural features and IgE binding competition with homologous allergens. Mol Immunol. 2019;114:189-95.
  26. Ehrenberg AE, Klingebiel C, Östling J, Larsson H, Mattsson L, Vitte J, et al. Characterization of a 7 kDa pollen allergen belonging to the gibberellin‐regulated protein family from three Cupressaceae species. Clinical & Experimental Allergy. 2020.
  27. Asam C, Hofer H, Wolf M, Aglas L, Wallner M. Tree pollen allergens—an update from a molecular perspective. Allergy. 2015;70(10):1201-11.
  28. Aceituno E, Del Pozo V, Mínguez A, Arrieta I, Cortegano I, Cárdaba B, et al. Molecular cloning of major allergen from Cupressus arizonica pollen: Cup a 1. Clin Exp Allergy. 2000;30(12):1750-8.
  29. Bar Dayan Y, Keynan N, WAISEL Y, PICK AI, TAMIR R. Podocarpus gracilior and Callitris verrucosa—newly identified allergens that crossreact with Cupressus sempervirens. Clinical & Experimental Allergy. 1995;25(5):456-60.
  30. Gonzalez E, Villalba M, Rodriguez R. Allergenic cross‐reactivity of olive pollen. Allergy. 2000;55(7):658-63.
  31. Pham N, Baldo B. Allergenic relationship between taxonomically diverse pollens. Clinical & Experimental Allergy. 1995;25(7):599-606.
  32. Hugues B, Didierlaurent A, Charpin D. Cross-reactivity between cypress pollen and peach: a report of seven cases. Allergy. 2006;61(10):1241-3.