Whole Allergen
w3 Giant Ragweed
w3 Giant Ragweed Scientific Information
Type:
Whole Allergen
Display Name:
Giant Ragweed
Allergen code:
w3
Family:
Asteraceae (Compositae)
Species:
A. trifida
Route of Exposure:
Inhalation
Source Material:
Giant ragweed pollen
Latin Name:
Ambrosia trifida
Other Names:
Great ragweed, giant ragweed, tall ragweed, blood ragweed, Texan great ragweed
WHO/ICD-11 code:
XM1X29
(ICD-11 is currently under implementation by WHO and the ICD-11 codes displayed in the encyclopedia may not yet be available in all countries)
Summary
Giant ragweed (Ambrosia trifida) is an herbaceous annual weed, native to North America and introduced to many countries in Europe and Asia. It is associated with allergic rhinitis, conjunctivitis and asthma as well as contact dermatitis. Giant ragweed shows high levels of cross-reactivity with other ragweeds (Ambrosia species) and mugwort (Artemisia species).
Allergen
Nature
Giant ragweed (Ambrosia trifida) is an herbaceous annual weed native to North America and Mexico. It has been introduced to many countries in Europe and Asia and is the most prevalent ragweed species in northern China (1-3). Giant ragweed is often one of the earliest plants to emerge during the spring and is mainly wind-pollinated. It is estimated that a single plant can produce up to 10 million pollen grains per day and one billion pollen grains in its lifetime (1, 3).
The ragweed (Ambrosia spp) pollen season has become significantly longer over the last 20 years, possibly due to climate change influences on plant distribution, flowering season, pollen load and allergenicity. Weed pollen and grass pollen seasons overlap in most parts of Europe, as do ragweed and mugwort pollen seasons (4).
Taxonomy      
Taxonomic tree of Ambrosia trifida (1)  
Domain
Eukaryota
Kingdom
Plantae
Phylum
Spermatophyta
Subphylum
Angiospermae
Class
Dicotyledonae
Family
Asteraceae
Genus
Ambrosia.
Tissue
Weed pollen
Epidemiology
Worldwide distribution 
Ragweed pollen is an important allergen source: Ambrosia species (of which short ragweed, giant ragweed and western ragweed are the most widespread) are major elicitors of pollen allergy in North America. In a study of 50 Chinese patients (aged 8–54 years) with weed pollen-induced allergic rhinitis, 34 % had specific IgE to giant ragweed allergen, Amb t (2)
Risk factors 
Climate change modelling suggests there will be an impact on the giant ragweed distribution area, although whether this will potentially increase or decrease depends on the climate scenario (5).
Environmental Characteristics
Worldwide distribution
Giant ragweed is an aggressive weed that prefers disturbed ground, including cultivated land, pasture, roadsides, riverbanks and grasslands, especially flood plains. This enables it to dominate the growing season, leading to poor yields and reduced biodiversity of native species (1).
Route of Exposure
Main 
The main route of exposure is inhalation.
Clinical Relevance
Giant ragweed pollen causes seasonal allergic rhinitis, asthma and conjunctivitis in sensitized patients as well as contact dermatitis (3, 6).
Asthma
Giant ragweed pollen can rupture osmotically in the atmosphere, producing sub-pollen particles that penetrate deeper into the respiratory tract than intact pollen grains, potentially triggering severe asthma 
Prevention and Therapy
Allergen immunotherapy
Allergenic cross-reactivity exists between Amb a (short or common ragweed), Amb p (western ragweed) and Amb t (giant ragweed), and given the effective Amb b inhibition of Amb p and Amb t suggests that an Amb a-based, single-species allergen immunotherapy may be successful in patients allergic to diverse ragweed pollens, although species-specific immunotherapy may be required given the allergen diversity (7, 8)
Molecular Aspects
Allergenic molecules    
Allergen
Class
Amb t 5
4.4 kDa basic protein
Amb t 5.0101
Isoform of Amb t 5
Amb t 8
Profilin
Amb t 5 (a secreted basic protein) is an identified allergen from giant ragweed. Amb t 5 is homologous to Amb a 5 from short or common ragweed (A. artemisiifolia), although their sequence homology is relatively low at 45 % (6). In a proteomic study of  other proteins were identified including some pectate lyases (Amb a 1.2, 1.4, and Amb a 2) as well as a profilin and profilin-3 (9).
Cross-reactivity
The presence of homologous allergens leads to extensive IgE cross-reactivity between the pollen extracts Amb t from giant ragweed, Amb p from western ragweed (A. psilostachya) and Amb a from short or common ragweed (A. artemisiifolia) (6, 8)
A high degree of IgE cross-reactivity is seen among different species of ragweed (Ambrosia spp) and mugwort (Artemisia) and to other allergenic plants of the Asteraceae family. (4). 
Compiled By
Author: RubyDuke Communications
Reviewer: Dr. Michael Thorpe
Last reviewed:February 2022
References
  1. CABI. Ambrosia trifida (giant ragweed) Wallingford, UK2021 [cited 2021 14.12.21]. Available from: https://www.cabi.org/isc/datasheet/4693.
  2. Han D, Lai X, Gjesing B, Zhong N, Zhang L, Spangfort MD. The specific IgE reactivity pattern of weed pollen-induced allergic rhinitis patients. Acta Otolaryngol. 2011;131(5):533-8.
  3. LOINC. Giant Ragweed triggered histamine release [Units/volume] in Blood 2021 [cited 2021 14.12.21]. Available from: https://loinc.org/7649-7/.
  4. Matricardi PM, Kleine-Tebbe J, Hoffmann HJ, Valenta R, Hilger C, Hofmaier S, et al. EAACI Molecular Allergology User's Guide. Pediatr Allergy Immunol. 2016;27 Suppl 23:1-250.
  5. Rasmussen K, Thyrring J, Muscarella R, Borchsenius F. Climate-change-induced range shifts of three allergenic ragweeds (Ambrosia L.) in Europe and their potential impact on human health. PeerJ. 2017;5:e3104.
  6. Gadermaier G, Dedic A, Obermeyer G, Frank S, Himly M, Ferreira F. Biology of weed pollen allergens. Current Allergy and Asthma Reports. 2004;4(5):391-400.
  7. Christensen LH, Ipsen H, Nolte H, Maloney J, Nelson HS, Weber R, et al. Short ragweeds is highly cross-reactive with other ragweeds. Ann Allergy Asthma Immunol. 2015;115(6):490-5.e1.
  8. Würtzen PA, Hoof I, Christensen LH, Váczy Z, Henmar H, Salamanca G, et al. Diverse and highly cross-reactive T-cell responses in ragweed allergic patients independent of geographical region. Allergy. 2020;75(1):137-47.
  9. Barton JS, Schomacker R. Comparative protein profiles of the Ambrosia plants. Biochim Biophys Acta Proteins Proteom. 2017;1865(6):633-9.