    • See all product categories
  • Applications
    • Bioprocessing
  • Services
    • 360° CDMO and CRO Solutions
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Proteins & Peptides ›
          • Thrombopoietin Proteins
          Gibco
          Rat TPO (Thrombopoietin) Recombinant Protein, PeproTech®
          View all (8) Thrombopoietin proteins
          Tech Support
          Cite Rat TPO (Thrombopoietin) Recombinant Protein, PeproTech®
          Additional Information:
          {{banner.description}}
          • Testing Data (1)
          Rat TPO (Thrombopoietin) Protein in Bioactivity (Bioactivity)
          Group 53 Created with Sketch.
          Rat TPO (Thrombopoietin) Protein in Bioactivity (Bioactivity)
          Group 53 Created with Sketch.
          FIGURE: 1 / 1
          Rat TPO (Thrombopoietin) Protein (400-34-500UG) in Bioactivity
          Bioassay analysis of Rat TPO (Thrombopoietin) Recombinant Protein, PeproTech® (Product # 400-34-1MG). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          Rat TPO (Thrombopoietin) Protein in Bioactivity (Bioactivity)
          Product Details
          400-34-500UG
          Applications
          Tested Dilution
          Publications
          Control (Ctrl)
          Assay-dependent
          -
          Bioactivity (Bioactivity)
          Assay-dependent
          -
          Product Specifications
          Species
          Rat
          Expression System
          E. coli
          Amino acid sequence
          SPVPPACDPR LLNKLLRDSY LLHSRLSQCP DVNPLSIPVL LPAVDFSLGE WKTQTEQSKA QDILGAVSLL LEGVMAARGQ LEPSCLSSLL GQLSGQVRLL LGALQGLLGT QLPPQGRTTA HKDPSALFLS LQQLLRGKVR FLLLVEGPAL CVRRTLPTTA VPSRTSQLLT LNKF
          Molecular weight
          18.7 kDa
          Class
          Recombinant
          Type
          Protein
          Purity
          ≥ 98% by SDS-PAGE gel and HPLC analyses.
          Endotoxin concentration
          <1 EU/µg
          Activity
          Determined by its ability to stimulate the proliferation of human MO7e cells. The expected ED50 is ≤ 0.2 ng/ml, corresponding to a specific activity of ≥ 5 x 10^6 units/mg.
          Conjugate
          Unconjugated Unconjugated Unconjugated
          Form
          Lyophilized
          Amount
          500 µg
          Purification
          purified
          Contains
          no preservative
          Storage conditions
          -20°C
          Shipping conditions
          Ambient
          Product Specific Information
          Recombinant Rat TPO is a fully biologically active 174 amino acid polypeptide (18.7 kDa), which contains the erythropoietin-like domain of the full length TPO protein.
          This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis
          Target Information
          Thrombopoietin (TPO, THPO, MPL ligand, megakaryocyte colony-stimulating factor) is responsible for the generation of platelets through its action on hematopoietic cells committed to the megakaryocyte lineage. Binding of thrombopoietin to its receptor, MPL (CD110), initiates a signaling cascade via JAK/STAT, MAPK, and PI3K/AKT signaling pathways that leads to the proliferation and differentiation of megakaryocytes and, ultimately, to the production of platelets (thrombopoiesis). Thrombopoietin is constitutively produced by the liver and has a predicted molecular weight of 38 kDa.
          Bioinformatics
          Gene Aliases: Thpo
          View more View less
          UniProt ID: (Rat) P49745
          View more View less
          Entrez Gene ID: (Rat) 81811
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          Your items have has been added!
          Host server : magellan-search-blue-76c4fffd86-q2vkv:80/100.66.78.131:80.
          git-commit: bdc067a1e9e92d056899a2dda64af631988aa8a1
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.54.0-Offline