Search
Search

SUT2| Gene Symbol: SUT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_126341.3 | NP_178389.1 | 1 - 2 | 588 | 83 | |
| NM_201675.2 | NP_973404.1 | 1 - 2 | 588 | 83 | ||
| GenBank mRNA | AJ289165.1 | - | 1 - 2 | 265 | 83 | |
| AK226970.1 | - | 1 - 2 | 392 | 83 | ||
Set Membership: |
|