Search
Search

GC1| Gene Symbol: GC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001342841.1 | NP_001318487.1 | 7 - 8 | 1199 | 82 | |
| NM_001342843.1 | NP_001331580.1 | 6 - 7 | 1134 | 82 | ||
| NM_001342844.1 | NP_001331579.1 | 6 - 7 | 1316 | 82 | ||
| NM_001342845.1 | NP_001331578.1 | 5 - 6 | 535 | 82 | ||
| NM_120675.5 | NP_568159.1 | 7 - 8 | 1028 | 82 | ||
| GenBank mRNA | AK229272.1 | - | 7 - 8 | 852 | 82 | |
| AY118140.1 | - | 6 - 7 | 632 | 82 | ||
| BT024499.1 | - | 6 - 7 | 632 | 82 | ||
Set Membership: |
|