Search
Search

RS2Z32| Gene Symbol: RS2Z32 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001339619.1 | NP_001326575.1 | 4 - 5 | 1091 | 81 | |
| NM_115210.6 | NP_190918.3 | 5 - 6 | 603 | 81 | ||
| NM_180684.4 | NP_851015.1 | 5 - 6 | 815 | 81 | ||
| GenBank mRNA | AK316697.1 | - | 5 - 6 | 527 | 81 | |
| AY095996.1 | - | 5 - 6 | 744 | 81 | ||
| BT001034.1 | - | 3 - 4 | 317 | 81 | ||
Set Membership: |
|