Search
Search

PRN| Gene Symbol: PRN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_115784.3 | NP_191481.1 | 5 - 6 | 783 | 147 | |
| GenBank mRNA | AF353716.1 | - | 5 - 6 | 461 | 147 | |
| BT026056.1 | - | 5 - 6 | 390 | 147 | ||
Set Membership: |
|