Search
Search

APG5| Gene Symbol: APG5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_121735.5 | NP_197231.1 | 1 - 2 | 228 | 86 | |
| GenBank mRNA | AK117572.1 | - | 1 - 2 | 114 | 86 | |
| BT005100.1 | - | 1 - 2 | 102 | 86 | ||
Set Membership: |
|