Search
Search

FC2| Gene Symbol: FC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001341698.1 | NP_001329102.1 | 8 - 9 | 925 | 65 | |
| NM_001341699.1 | NP_001329107.1 | 10 - 11 | 1418 | 65 | ||
| NM_001341700.1 | NP_001329103.1 | 10 - 11 | 1396 | 65 | ||
| NM_001341701.1 | NP_001329104.1 | 8 - 9 | 2169 | 65 | ||
| NM_001341702.1 | NP_001329105.1 | 9 - 10 | 2076 | 65 | ||
| NM_001341703.1 | NP_001329106.1 | 10 - 11 | 1931 | 65 | ||
| NM_001341704.1 | NP_001329108.1 | 7 - 8 | 1912 | 65 | ||
| NM_001341705.1 | NP_001329109.1 | 10 - 11 | 1459 | 65 | ||
| NM_118607.3 | NP_194205.1 | 11 - 12 | 1359 | 65 | ||
| NM_202881.2 | NP_974610.1 | 10 - 11 | 1169 | 65 | ||
| GenBank mRNA | AK227055.1 | - | 11 - 12 | 1231 | 65 | |
| BT025991.1 | - | 11 - 12 | 1146 | 65 | ||
| D45353.1 | - | 11 - 12 | 1266 | 65 | ||
| U16177.1 | - | 11 - 12 | 1189 | 65 | ||
Set Membership: |
|