Search
Search

KNAT5| Gene Symbol: KNAT5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001342125.1 | NP_001328629.1 | 2 - 3 | 794 | 119 | |
| NM_119356.4 | NP_194932.1 | 2 - 3 | 799 | 119 | ||
| GenBank mRNA | AF306661.1 | - | 2 - 3 | 679 | 119 | |
| BT026033.1 | - | 2 - 3 | 661 | 119 | ||
| D43962.1 | - | 2 - 3 | 705 | 119 | ||
Set Membership: |
|