Search
Search

HDR| Gene Symbol: HDR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_119600.4 | NP_567965.1 | 1 - 2 | 381 | 70 | |
| GenBank mRNA | AY042877.1 | - | 1 - 2 | 351 | 70 | |
| AY081454.1 | - | 1 - 2 | 275 | 70 | ||
| AY168881.1 | - | 1 - 2 | 299 | 70 | ||
Set Membership: |
|