Search
Search

GOS12| Gene Symbol: GOS12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_130083.5 | NP_182045.1 | 1 - 2 | 438 | 57 | |
| GenBank mRNA | AF357529.1 | - | 1 - 2 | 187 | 57 | |
| BT024623.1 | - | 1 - 2 | 132 | 57 | ||
Set Membership: |
|