Search
Search

UXS4| Gene Symbol: UXS4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001202843.1 | NP_001189772.1 | 3 - 4 | 954 | 83 | |
| NM_130333.4 | NP_182287.1 | 3 - 4 | 1066 | 83 | ||
| GenBank mRNA | AK316734.1 | - | 3 - 4 | 955 | 83 | |
| AK318797.1 | - | 3 - 4 | 953 | 83 | ||
| BT022016.1 | - | 3 - 4 | 857 | 83 | ||
Set Membership: |
|