Search
Search

CRCK1| Gene Symbol: CRCK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001345330.1 | NP_001318835.1 | 6 - 7 | 2025 | 69 | |
| NM_001345331.1 | NP_001332082.1 | 7 - 8 | 1470 | 69 | ||
| NM_001345332.1 | NP_001332083.1 | 6 - 7 | 1422 | 69 | ||
| NM_125284.4 | NP_200702.2 | 5 - 6 | 1500 | 69 | ||
| GenBank mRNA | BT015092.1 | - | 5 - 6 | 1171 | 69 | |
| BT015906.1 | - | 5 - 6 | 1098 | 69 | ||
Set Membership: |
|