Search
Search

PRH75| Gene Symbol: PRH75 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_125613.3 | NP_201025.1 | 1 - 2 | 597 | 86 | |
| GenBank mRNA | AY056137.1 | - | 1 - 2 | 523 | 86 | |
| BT008581.1 | - | 1 - 2 | 432 | 86 | ||
Set Membership: |
|