Search
Search

AAO3| Gene Symbol: AAO3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001084497.1 | NP_001077966.1 | 6 - 7 | 3593 | 149 | |
| NM_001336104.1 | NP_001324268.1 | 6 - 7 | 3520 | 149 | ||
| NM_001336105.1 | NP_001324270.1 | 6 - 7 | 3615 | 149 | ||
| NM_001336106.1 | NP_001324271.1 | 6 - 7 | 3515 | 149 | ||
| NM_001336107.1 | NP_001324269.1 | 6 - 7 | 3493 | 149 | ||
| NM_128273.3 | NP_180283.1 | 6 - 7 | 3620 | 149 | ||
Set Membership: |
|