Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ?
Create Account
Applications
Real-Time PCR
Oligos, Primers, Probes and Genes
Cloning
Protein Biology
Cell Culture and Transfection
Cell and Gene Therapy
Cell Analysis
Mass Spectrometry
Chromatography
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Online Order
Custom Primers & TaqMan Probes
miRNA Mimics & Inhibitors
Stealth RNAi / siRNA
Silencer Select siRNAs
Custom DNA Oligos
GeneArt Services
GeneArt Strings DNA Fragments
TrueGuide CRISPR gRNA
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
See all online orderable products
Services
Custom Services
Instrument Qualification Services
Technical Services
Pipette Services
Sample Request
Instrument Maintenance Services
Repairs and Relocation Services
OEM and Licensing Services
Events, Seminars, Training
Unity Lab Services
See all services
Support
Catalogs
Application Notes
Safety Data Sheets
Legal and Regulatory
Certificates of Analysis Search
On-demand videos
FAQs
Learning Centers
Technical Support Centers
See all help and support topics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign In
Sign In
Don't have an account ?
Create Account
Connect Your Lab
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Home
›
Search Tool
›
Search Results
› At02280471_g1
See other ATB' DELTA GE Assays ›
Gene Symbol:
ATB' DELTA
Gene Name:
serine/threonine protein phosphatase 2A 55 kDa regulatory subunit B prime delta
Gene Aliases:
AT3G26030, MPE11.23, serine/threonine protein phosphatase 2A 55 kDa regulatory subunit B prime delta
Chromosome Location:
Chr.3: 9516953 - 9519563 on Build TAIR10.1
Species:
Arabidopsis
Species Specific ID (Flybase ID):
-
Assay ID
At02280471_g1
Best Coverage
Size
FAM (900 nM Primer) | XS: 75 rxns
FAM (900 nM Primer) | S: 360 rxns
FAM (900 nM Primer) | M: 750 rxns
FAM (900 nM Primer) | L: 2,900 rxns
FAM PL (350 nM Primer Limited) | S: 360 rxns
FAM PL (350 nM Primer Limited) | M: 750 rxns
FAM PL (350 nM Primer Limited) | L: 2,900 rxns
VIC (900 nM Primer) | S: 360 rxns
VIC (900 nM Primer) | M: 750 rxns
VIC (900 nM Primer) | L: 2,900 rxns
VIC PL (150 nM Primer Limited) | S: 360 rxns
VIC PL (150 nM Primer Limited) | M: 750 rxns
VIC PL (150 nM Primer Limited) | L: 2,900 rxns
ABY PL (350 nM Primer Limited) | S: 360 rxns
ABY PL (350 nM Primer Limited) | M: 750 rxns
ABY PL (350 nM Primer Limited) | L: 2,900 rxns
JUN PL (350 nM Primer Limited) | S: 360 rxns
JUN PL (350 nM Primer Limited) | M: 750 rxns
JUN PL (350 nM Primer Limited) | L: 2,900 rxns
CYN5 PL (350 nM Primer Limited) | S: 360 rxns
CYN5 PL (350 nM Primer Limited) | M: 750 rxns
CYN5 PL (350 nM Primer Limited) | L: 2,900 rxns
Availability
Made To Order
Catalog #
4351372
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Gene Symbol: ATB' DELTA
Interrogated Sequence
Translated Protein
Exon Boundary
Assay Location
IMAGE Clone ID
Amplicon Length
RefSeq
NM_113507.3
NP_189232.1
1 - 2
1917
138
GenBank mRNA
AF062396.1
-
1 - 2
1537
138
AY091037.1
-
1 - 2
1557
138
AY117349.1
-
1 - 2
1129
138
Back To Top
More Information
Set Membership:
Amplicon length greater than or equal to 101
3' Most
Detect Genomic DNA
Recommended Assay
Arabidopsis thaliana
Probe spans exons
Panther Classification:
Molecular Function -
protein phosphatase
Gene Ontology Categories:
Function(s)
Process(es)
signal transduction
protein binding
protein phosphatase regulator activity
Back To Top
Related Products
TaqMan
®
Fast Advanced Master Mix
SuperScript
®
VILO
™
cDNA Synthesis Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-7fb9f846c9-l8rg4:80/100.66.76.64:80.
git-commit: 8207b5afa5c305bd40e0a4f0b9b53fd46f2a8829
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.59.1-2026.09.91-1.0