Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: AAR3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001338993.1 | NP_001326021.1 | 5 - 6 | 540 | 111 | |
NM_113818.4 | NP_189539.1 | 5 - 6 | 633 | 111 | ||
GenBank mRNA | AK118926.1 | - | 5 - 6 | 538 | 111 | |
BT005690.1 | - | 5 - 6 | 399 | 111 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |