Search
Search

BR6OX2| Gene Symbol: BR6OX2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_113917.4 | NP_566852.1 | 2 - 3 | 661 | 147 | |
| GenBank mRNA | AB087801.1 | - | 2 - 3 | 559 | 147 | |
| AY052655.1 | - | 2 - 3 | 611 | 147 | ||
| AY063728.1 | - | 2 - 3 | 520 | 147 | ||
Set Membership: |
|