Search
Search

BCAT3| Gene Symbol: BCAT3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001125330.1 | NP_001118802.1 | 8 - 9 | 1123 | 82 | |
| NM_114828.6 | NP_566923.1 | 8 - 9 | 1236 | 82 | ||
| GenBank mRNA | AF446355.1 | - | 8 - 9 | 1101 | 82 | |
| AJ276124.1 | - | 8 - 9 | 1103 | 82 | ||
| AY097417.1 | - | 8 - 9 | 1010 | 82 | ||
Set Membership: |
|