Search
Search

MPK5| Gene Symbol: MPK5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_117204.7 | NP_567378.4 | 5 - 6 | 1170 | 83 | |
| GenBank mRNA | AK176361.1 | - | 5 - 6 | 1043 | 83 | |
| GU989202.1 | - | 5 - 6 | 959 | 83 | ||
Set Membership: |
|