Search
Search

ARD4| Gene Symbol: ARD4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_123752.4 | NP_568630.1 | 4 - 5 | 680 | 75 | |
| GenBank mRNA | BT000432.1 | - | 4 - 5 | 475 | 75 | |
| BT006566.1 | - | 3 - 4 | 439 | 75 | ||
Set Membership: |
|