Search
Search

CESA4| Gene Symbol: CESA4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001344528.1 | NP_001332727.1 | 4 - 5 | 984 | 106 | |
| NM_123770.4 | NP_199216.2 | 5 - 6 | 897 | 106 | ||
| GenBank mRNA | AK228561.1 | - | 5 - 6 | 645 | 106 | |
| BT005710.1 | - | 5 - 6 | 645 | 106 | ||
| BT006111.1 | - | 5 - 6 | 574 | 106 | ||
| KJ676989.1 | - | 5 - 6 | 574 | 106 | ||
Set Membership: |
|