Search
Search

C22H3orf14| Gene Symbol: C22H3orf14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001046510.1 | NP_001039975.1 | 1 - 2 | 741 | 110 | |
| GenBank mRNA | BC113216.1 | - | 1 - 2 | 741 | 110 | |
Set Membership: |
|