Search
Search

csk-1| Gene Symbol: csk-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001306233.3 | NP_001293162.1 | 12 - 13 | 1829 | 69 | |
| NM_001306234.3 | NP_001293163.1 | 1 - 2 | 86 | 69 | ||
| NM_001380379.1 | NP_001367885.1 | 11 - 12 | 1744 | 69 | ||
| GenBank mRNA | AB096875.1 | - | 11 - 12 | 1731 | 69 | |
Set Membership: |
|