Search
Search

aat-5| Gene Symbol: aat-5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_058601.6 | NP_491002.2 | 4 - 5 | 566 | 69 | |
| NR_131358.1 | 4 - 5 | 552 | 69 | |||
| NR_131359.1 | 4 - 5 | 552 | 69 | |||
| NR_131780.1 | 4 - 5 | 552 | 69 | |||
Set Membership: |
|