Search
Search

spe-9| Gene Symbol: spe-9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001136278.3 | NP_001129750.1 | 8 - 9 | 1001 | 65 | |
| NM_060554.5 | NP_492955.1 | 15 - 16 | 1866 | 65 | ||
| GenBank mRNA | AF049327.1 | - | 15 - 16 | 1844 | 65 | |
Set Membership: |
|