Search
Search

dct-6| Gene Symbol: dct-6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001038268.4 | NP_001033357.2 | 2 - 3 | 134 | 105 | |
| NM_001038269.5 | NP_001033358.2 | 2 - 3 | 134 | 105 | ||
Set Membership: |
|