Search
Search

pqn-80| Gene Symbol: pqn-80 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001268248.4 | NP_001255177.1 | 17 - 18 | 3539 | 90 | |
| NM_001268249.2 | NP_001255178.1 | 16 - 17 | 3456 | 90 | ||
| NM_001312958.3 | NP_001299887.1 | 17 - 18 | 3555 | 90 | ||
| NM_001312959.3 | NP_001299888.1 | 16 - 17 | 3458 | 90 | ||
| NM_001312960.3 | NP_001299889.1 | 14 - 15 | 2969 | 90 | ||
| NM_001312961.3 | NP_001299890.1 | 13 - 14 | 2879 | 90 | ||
| NM_001312962.3 | NP_001299891.1 | 14 - 15 | 2960 | 90 | ||
| NM_001312963.3 | NP_001299892.1 | 13 - 14 | 2870 | 90 | ||
| NM_001312964.3 | NP_001299893.1 | 13 - 14 | 2399 | 90 | ||
| NM_001312965.3 | NP_001299894.1 | 12 - 13 | 2309 | 90 | ||
| NM_001312966.3 | NP_001299895.1 | 13 - 14 | 2390 | 90 | ||
| NM_001312967.3 | NP_001299896.1 | 12 - 13 | 2300 | 90 | ||
| NM_001312968.3 | NP_001299897.1 | 4 - 5 | 971 | 90 | ||
| NM_001312969.3 | NP_001299898.1 | 3 - 4 | 881 | 90 | ||
| NM_001312970.3 | NP_001299899.1 | 2 - 3 | 362 | 90 | ||
Set Membership: |
|