Search
Search

tax-6| Gene Symbol: tax-6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001026121.2 | NP_001021292.1 | 6 - 7 | 1108 | 64 | |
| NM_001083189.5 | NP_001076658.2 | 6 - 7 | 1108 | 64 | ||
| NM_001380448.1 | NP_001367052.1 | 5 - 6 | 856 | 64 | ||
Set Membership: |
|