Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: unc-83 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001083280.3 | NP_001076749.1 | 15 - 16 | 2989 | 99 | |
NM_001373040.1 | NP_001360140.1 | 15 - 16 | 2989 | 99 | ||
NM_001383319.1 | 15 - 16 | 2967 | 99 | |||
GenBank mRNA | AF338767.1 | - | 15 - 16 | 2976 | 99 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |