Search
Search
| Gene Symbol: MORN4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005637593.3 | XP_005637650.1 | 5 - 6 | 553 | 141 | |
| XM_005637596.3 | XP_005637653.1 | 3 - 4 | 363 | 141 | ||
| XM_014108995.2 | XP_013964470.1 | 5 - 6 | 579 | 141 | ||
| XM_014108996.2 | XP_013964471.1 | 5 - 6 | 624 | 141 | ||
| XM_014108997.2 | XP_013964472.1 | 6 - 7 | 662 | 141 | ||
| XM_014108999.2 | XP_013964474.1 | 4 - 5 | 486 | 141 | ||
| XM_022411644.1 | XP_022267352.1 | 3 - 4 | 625 | 141 | ||
| XM_022411645.1 | XP_022267353.1 | 6 - 7 | 696 | 141 | ||
| XM_022411646.1 | XP_022267354.1 | 6 - 7 | 741 | 141 | ||
| XM_022411647.1 | XP_022267355.1 | 6 - 7 | 636 | 141 | ||
| XM_022411648.1 | XP_022267356.1 | 5 - 6 | 598 | 141 | ||
| XM_534983.5 | XP_534983.2 | 4 - 5 | 460 | 141 | ||
Set Membership: |
|