Search
Search
| Gene Symbol: NCBP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005639604.3 | XP_005639661.1 | 2 - 3 | 324 | 124 | |
| XM_014110197.2 | XP_013965672.1 | 2 - 3 | 324 | 124 | ||
| XM_022413826.1 | XP_022269534.1 | 2 - 3 | 324 | 124 | ||
Set Membership: |
|