Search
Search
| Gene Symbol: MYLK4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005639977.3 | XP_005640034.1 | 2 - 3 | 526 | 117 | |
| XM_022414393.1 | XP_022270101.1 | 2 - 3 | 383 | 117 | ||
| XM_545312.6 | XP_545312.4 | 2 - 3 | 383 | 117 | ||
Set Membership: |
|