Search
Search
| Gene Symbol: CALML6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005620406.3 | XP_005620463.1 | 506 | 63 | ||
| XM_005620410.3 | XP_005620467.1 | 367 | 63 | |||
| XM_022418914.1 | XP_022274622.1 | 454 | 63 | |||
| XM_022418915.1 | XP_022274623.1 | 442 | 63 | |||
| XM_022418916.1 | XP_022274624.1 | 499 | 63 | |||
| XM_022418917.1 | XP_022274625.1 | 495 | 63 | |||
| XM_022418918.1 | XP_022274626.1 | 467 | 63 | |||
| XM_022418919.1 | XP_022274627.1 | 419 | 63 | |||
Set Membership: |
|