Search
Search

Idua| Gene Symbol: Idua | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007642986.1 | XP_007641176.1 | 10 - 11 | 1964 | 58 | |
| GenBank mRNA | JI874305.1 | - | 2 - 3 | 58 | ||
| JP054043.1 | - | 10 - 11 | 1486 | 58 | ||
Set Membership: |
|