Search
Search
| Gene Symbol: DIMT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001285600.2 | NP_001272529.1 | 4 - 5 | 355 | 74 | |
| GenBank mRNA | JN942971.1 | - | 4 - 5 | 355 | 74 | |
| JT749293.1 | - | 4 - 5 | 432 | 74 | ||
Set Membership: |
|