Search
Search
| Gene Symbol: HCN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018050508.1 | XP_017905997.1 | 3 - 4 | 1279 | 58 | |
| GenBank mRNA | JO632303.2 | - | 5 - 6 | 2115 | 58 | |
| JT639651.1 | - | 3 - 4 | 738 | 58 | ||
Set Membership: |
|