Search
Search

ham| Gene Symbol: ham | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001259152.1 | NP_001246081.1 | 4 - 5 | 1209 | 102 | |
| NM_001273614.1 | NP_001260543.1 | 4 - 5 | 1541 | 102 | ||
| NM_001273616.1 | NP_001260545.2 | 3 - 4 | 1078 | 102 | ||
| NM_165262.3 | NP_724130.4 | 4 - 5 | 1210 | 102 | ||
| GenBank mRNA | AY166585.1 | - | 3 - 4 | 560 | 102 | |
Set Membership: |
|