Search
Search

stan| Gene Symbol: stan | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001169642.1 | NP_001163113.1 | 1 - 2 | 2119 | 91 | |
| NM_001201974.2 | NP_001188903.1 | 3 - 4 | 2371 | 91 | ||
| NM_001259328.2 | NP_001246257.1 | 3 - 4 | 2371 | 91 | ||
| NM_001259329.2 | NP_001246258.1 | 3 - 4 | 2371 | 91 | ||
| NM_001273942.1 | NP_001260871.1 | 3 - 4 | 2371 | 91 | ||
| NM_001299362.1 | NP_001286291.1 | 3 - 4 | 2371 | 91 | ||
| NM_165794.4 | NP_724962.3 | 1 - 2 | 2119 | 91 | ||
Set Membership: |
|