Search
Search

Nf-YB| Gene Symbol: Nf-YB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001299160.1 | NP_001286089.1 | 1 - 2 | 169 | 73 | |
| NM_136153.4 | NP_609997.1 | 1 - 2 | 169 | 73 | ||
| GenBank mRNA | AY070968.1 | - | 1 - 2 | 135 | 73 | |
| AY094930.1 | - | 1 - 2 | 158 | 73 | ||
Set Membership: |
|