Search
Search

| Gene Symbol: rg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001042796.3 | NP_001036261.3 | 5 - 6 | 1362 | 62 | |
| NM_001144685.2 | NP_001138157.2 | 5 - 6 | 1674 | 62 | ||
| NM_001144686.2 | NP_001138158.2 | 5 - 6 | 1674 | 62 | ||
| NM_001144687.2 | NP_001138159.2 | 5 - 6 | 1674 | 62 | ||
| NM_001297949.1 | NP_001284878.1 | 8 - 9 | 5691 | 62 | ||
| NM_001297950.1 | NP_001284879.1 | 5 - 6 | 1674 | 62 | ||
| NM_167028.3 | NP_726978.2 | 6 - 7 | 3035 | 62 | ||
| Gene Symbol: CG15465 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001297948.1 | NP_001284877.1 | 8 - 9 | 5691 | 62 | |
| NM_131978.3 | NP_572206.1 | 6 - 7 | 3035 | 62 | ||
Set Membership: |
|