Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: ATP7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001272537.2 | NP_001259466.1 | 3 - 4 | 3340 | 57 | |
NM_132528.3 | NP_572756.3 | 3 - 4 | 3340 | 57 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Drosophila melanogaster |