Search
Search

Src64B| Gene Symbol: Src64B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014561.2 | NP_001014561.1 | 5 - 6 | 1770 | 62 | |
| NM_001014562.3 | NP_001014562.1 | 4 - 5 | 1522 | 62 | ||
| NM_001014563.3 | NP_001014563.1 | 6 - 7 | 1911 | 62 | ||
| NM_001014564.3 | NP_001014564.1 | 6 - 7 | 1672 | 62 | ||
| NM_001202122.1 | NP_001189051.1 | 5 - 6 | 1465 | 62 | ||
| NM_001259699.2 | NP_001246628.1 | 6 - 7 | 1672 | 62 | ||
| NM_001259700.2 | NP_001246629.1 | 6 - 7 | 1672 | 62 | ||
| NM_080195.4 | NP_524934.2 | 6 - 7 | 1737 | 62 | ||
| GenBank mRNA | AY051781.1 | - | 6 - 7 | 1714 | 62 | |
Set Membership: |
|