Search
Search

sns| Gene Symbol: sns | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001043067.2 | NP_001036532.1 | 8 - 9 | 2014 | 89 | |
| NM_001299284.1 | NP_001286213.1 | 8 - 9 | 2014 | 89 | ||
| NM_176106.2 | NP_788286.1 | 8 - 9 | 2014 | 89 | ||
Set Membership: |
|