Search
Search

Pkn| Gene Symbol: Pkn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001201966.2 | NP_001188895.1 | 7 - 8 | 3355 | 76 | |
| NM_001201967.1 | NP_001188896.1 | 5 - 6 | 2970 | 76 | ||
| NM_001201968.3 | NP_001188897.3 | 6 - 7 | 2875 | 76 | ||
| NM_001259291.2 | NP_001246220.1 | 6 - 7 | 2816 | 76 | ||
| NM_001259292.2 | NP_001246221.1 | 6 - 7 | 2947 | 76 | ||
| NM_001299312.1 | NP_001286241.1 | 7 - 8 | 3467 | 76 | ||
| NM_176114.3 | NP_788294.1 | 2 - 3 | 476 | 76 | ||
| GenBank mRNA | BT133361.1 | - | 2 - 3 | 485 | 76 | |
Set Membership: |
|