Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Dm02134477_m1
          See other HTH GE Assays ›
          Gene Symbol:
          hth
          Gene Name:
          homothorax
          Gene Aliases:
          1323/07, 1422/04, CG17117, Dm-HTH, Dmel\CG17117, Dmel_CG17117, HTH, Hth, MEIS1, Meis1, P53, anon-EST:Liang-2.13, clone 2.13, dtl, hth1, hth2, l(3)05745, l(3)86Ca
          Chromosome Location:
          Chr.3R: 10507561 - 10639568 on Build Release_6_plus_ISO1_MT
          Species:
          Drosophila melanogaster
          Species Specific ID (Flybase ID):
          FBgn0001235
          Assay ID Dm02134477_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: hth
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001014613.3 NP_001014613.2 3 - 4 2045 73
          NM_001031999.2 NP_001027170.1 2 - 3 1576 73
          NM_057228.5 NP_476576.1 2 - 3 1576 73
          NM_057229.5 NP_476577.3 2 - 3 1576 73
          NM_057230.6 NP_476578.3 2 - 3 1576 73
          GenBank mRNA AF032865.1 - 2 - 3 670 73
          AF035825.1 - 2 - 3 1552 73
          AF036584.1 - 2 - 3 757 73
          BT024210.1 - 2 - 3 1395 73
          BT149862.1 - 3 - 4 1141 73

          Back To Top

          More Information


          Set Membership:

          Amplicon length between 71 and 85 Recommended Assay Drosophila melanogaster

          Panther Classification:

          Molecular Function -

          homeodomain transcription factor

          Gene Ontology Categories:

          Function(s) Process(es)

          eye development
          oenocyte differentiation
          compound eye photoreceptor fate commitment
          regulation of transcription from RNA polymerase II promoter
          specification of segmental identity, head
          specification of segmental identity, antennal segment
          brain development
          peripheral nervous system development
          salivary gland boundary specification
          imaginal disc-derived wing morphogenesis
          imaginal disc-derived leg morphogenesis
          somatic muscle development
          positive regulation of cell proliferation
          embryonic pattern specification
          organ morphogenesis
          proximal/distal pattern formation
          specification of organ identity
          protein localization to nucleus
          segmentation
          regulation of cell fate specification
          regulation of neuron differentiation
          positive regulation of transcription from RNA polymerase II promoter
          haltere morphogenesis
          compound eye development
          head morphogenesis
          Malpighian tubule development
          positive regulation of cell proliferation involved in compound eye morphogenesis
          RNA polymerase II core promoter proximal region sequence-specific DNA binding
          RNA polymerase II transcription factor activity, sequence-specific DNA binding
          core promoter proximal region sequence-specific DNA binding
          transcription coactivator binding
          transcriptional activator activity, RNA polymerase II transcription regulatory region sequence-specific binding
          DNA binding
          transcription factor activity, sequence-specific DNA binding
          protein binding
          transcription factor binding

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline