Search
Search

RhoGAP92B| Gene Symbol: RhoGAP92B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001300485.1 | NP_001287414.1 | 2 - 3 | 459 | 59 | |
| NM_001300487.1 | NP_001287416.1 | 1 - 2 | 444 | 59 | ||
| NM_001300488.1 | NP_001287417.1 | 2 - 3 | 459 | 59 | ||
| NM_142587.3 | NP_650844.1 | 2 - 3 | 459 | 59 | ||
| GenBank mRNA | BT044266.1 | - | 2 - 3 | 484 | 59 | |
Set Membership: |
|