Search
Search

sle| Gene Symbol: sle | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_169338.2 | NP_731472.1 | 2 - 3 | 287 | 95 | |
| NM_169339.2 | NP_731473.1 | 1 - 2 | 188 | 95 | ||
| GenBank mRNA | BT031151.1 | - | 2 - 3 | 226 | 95 | |
Set Membership: |
|