Search
Search

Fas1| Gene Symbol: Fas1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001104344.2 | NP_001097814.1 | 11 - 12 | 1716 | 91 | |
| NM_001144593.2 | NP_001138065.1 | 10 - 11 | 1624 | 91 | ||
| NM_001144594.2 | NP_001138066.1 | 10 - 11 | 1677 | 91 | ||
| NM_001275716.1 | NP_001262645.1 | 10 - 11 | 1677 | 91 | ||
| NM_169724.2 | NP_732164.1 | 12 - 13 | 1695 | 91 | ||
| NM_169725.2 | NP_732165.1 | 11 - 12 | 1633 | 91 | ||
| NM_169726.2 | NP_732166.1 | 10 - 11 | 1624 | 91 | ||
| GenBank mRNA | BT044209.1 | - | 10 - 11 | 1655 | 91 | |
| M20545.1 | - | 11 - 12 | 1612 | 91 | ||
Set Membership: |
|